Gene Transcripts

Gene Symbol
Phlpp2
Entrez Gene ID
244650
Gene Name
PH domain and leucine rich repeat protein phosphatase 2
Gene Aliases
AI481772, C130044A18Rik, Phlppl
Chromosome Location
Chr. 8: 109868519 - 109944671 on Build GRCm38
UniGene ID
Mm.23494
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001122594.2 NP_001116066.2 3 435
GenBank mRNA AK048260.1 - 3 392
AK048472.1 - 3 436
AK165088.1 - 3 497
BC117961.1 - 3 435 40098281
BC117962.1 - 3 427 40098282

View Full Product Details