Gene Transcripts

Gene Symbol
Ankk1
Entrez Gene ID
244859
Gene Name
ankyrin repeat and kinase domain containing 1
Gene Aliases
9930020N01Rik
Chromosome Location
Chr. 9: 49412955 - 49427791 on Build GRCm38
UniGene ID
Mm.119994
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172922.3 NP_766510.1 5 846
GenBank mRNA AK036872.1 - 5 847
BC145077.1 - 5 873
BC145078.1 - 5 836 9053626

View Full Product Details