Gene Transcripts

Gene Symbol
Fat2
Entrez Gene ID
245827
Gene Name
FAT tumor suppressor homolog 2 (Drosophila)
Gene Aliases
EMI2, Fath2, Gm523, mKIAA0811
Chromosome Location
Chr. 11: 55250609 - 55336621 on Build GRCm38
UniGene ID
Mm.339363
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001029988.2 NP_001025159.1 9 7483

View Full Product Details