Gene Transcripts

Gene Symbol
Atg9a
Entrez Gene ID
245860
Gene Name
autophagy related 9A
Gene Aliases
AU019532, Apg9l1, Atg9, Atg9l1
Chromosome Location
Chr. 1: 75180861 - 75192010 on Build GRCm38
UniGene ID
Mm.238266
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003917.4 NP_001003917.2 8 911
NM_001288612.1 NP_001275541.1 8 914
NM_001288613.1 NP_001275542.1 8 1159
GenBank mRNA AM085510.1 - 8 824
BC056488.1 - 8 1072
BC079884.1 - 8 859 6840050
BC082307.1 - 6 617

View Full Product Details