Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_139134.3 | NP_624360.2 | 5, 6 | 1150 | |
GenBank mRNA | AF311699.1 | - | 5, 6 | 973 | |
AK014255.1 | - | 5, 6 | 861 | ||
AK140910.1 | - | 5, 6 | 1151 | ||
BC117071.1 | - | 5, 6 | 821 | 40126390 | |
BC117073.1 | - | 5, 6 | 821 | 40126392 | |
BC144738.1 | - | 5, 6 | 806 | 9053286 |