Gene Transcripts

Gene Symbol
Defb9
Entrez Gene ID
246079
Gene Name
defensin beta 9
Gene Aliases
BD-9
Chromosome Location
Chr. 8: 21881713 - 21885434 on Build GRCm38
UniGene ID
Mm.171224
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139219.2 NP_631965.1 2 119
GenBank mRNA AJ437647.1 - 2 106
BC109177.2 - 2 102 40049218
BM936051.1 - 2 148

View Full Product Details