Gene Transcripts

Gene Symbol
MAPK13
Entrez Gene ID
5603
Gene Name
mitogen-activated protein kinase 13
Gene Aliases
MAPK 13, MAPK-13, PRKM13, SAPK4, p38delta
Chromosome Location
Chr. 6: 36130484 - 36144524 on Build GRCh38
UniGene ID
Hs.178695
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002754.4 NP_002745.1 4 497
NR_072996.1 4 497
GenBank mRNA AB209586.1 - 3 1455
AB451221.1 - 5 398
AB451344.1 - 5 398
AF004709.1 - 4 446
AF015256.1 - 4 458
AF092535.1 - 4 419
AF100546.1 - 4 400
AK308018.1 - 4 478
BC000433.2 - 4 439 2819932
BC001641.2 - 4 439 2819932
BC004428.1 - 4 439 2819932
BC085196.1 - 4 467 6107307
BM921729.1 - 4 497
BT007221.1 - 4 398
CR536490.1 - 4 398
CR541687.1 - 4 398
DB473379.1 - 4 461
DR006449.1 - 4 501
KU178281.1 - 4 398
KU178282.1 - 4 398
U93232.1 - 4 405
Y10488.1 - 4 420

View Full Product Details