Gene Transcripts

Gene Symbol
MAP2K5
Entrez Gene ID
5607
Gene Name
mitogen-activated protein kinase kinase 5
Gene Aliases
HsT17454, MAPKK5, MEK5, PRKMK5
Chromosome Location
Chr. 15: 67542606 - 67807117 on Build GRCh38
UniGene ID
Hs.114198
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001206804.1 NP_001193733.1 13 885
NM_002757.3 NP_002748.1 13 1469
NM_145160.2 NP_660143.1 13 1469
XM_011521787.2 XP_011520089.1 13 1546
XM_011521788.2 XP_011520090.1 13 1546
GenBank mRNA AK024766.1 - 2 63
AK057521.1 - 7 597
AK293459.1 - 13 885
AK315435.1 - 13 1016
BC008838.2 - 13 1126 4100679
BT006780.1 - 13 816
CR542229.1 - 13 816
U71087.1 - 13 877
U71088.2 - 13 821

View Full Product Details