Gene Transcripts

Gene Symbol
Olfr90
Entrez Gene ID
258469
Gene Name
olfactory receptor 90
Gene Aliases
MOR256-21, bM573K1.2
Chromosome Location
Chr. 17: 37085191 - 37086236 on Build GRCm38
UniGene ID
Mm.271392
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146477.2 NP_666688.2 1 588
GenBank mRNA BC119191.1 - 1 588

View Full Product Details