Gene Transcripts

Gene Symbol
Cdk8
Entrez Gene ID
264064
Gene Name
cyclin-dependent kinase 8
Gene Aliases
Chromosome Location
Chr. 5: 146230736 - 146302874 on Build GRCm38
UniGene ID
Mm.260576
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153599.3 NP_705827.2 4 353
GenBank mRNA AK182034.1 - 2 105
BI151839.1 - 4 353
CF723869.1 - 2 55
CF730982.1 - 4 271

View Full Product Details