Gene Transcripts

Gene Symbol
Rapgefl1
Entrez Gene ID
268480
Gene Name
Rap guanine nucleotide exchange factor (GEF)-like 1
Gene Aliases
B230105J10
Chromosome Location
Chr. 11: 98836785 - 98853006 on Build GRCm38
UniGene ID
Mm.340766
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080925.1 NP_001074394.1 5, 6 1316
GenBank mRNA BC080279.1 - 4, 5 384
BC130266.1 - 6, 7 592 40143028

View Full Product Details