Gene Transcripts

Gene Symbol
PSMD8
Entrez Gene ID
5714
Gene Name
proteasome 26S subunit, non-ATPase 8
Gene Aliases
HEL-S-91n, HIP6, HYPF, Nin1p, Rpn12, S14, p31
Chromosome Location
Chr. 19: 38374550 - 38383824 on Build GRCh38
UniGene ID
Hs.78466
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002812.4 NP_002803.2 3 578
GenBank mRNA AK301771.1 - 3 589
AK308809.1 - 3 667
BC001164.1 - 3 329 3528096
BC065006.1 - 3 317 6055235
BI560993.1 - 3 562
BQ684248.1 - 3 341
BT019688.1 - 3 247
D38047.1 - 3 317
EU668354.1 - 3 578

View Full Product Details