Gene Transcripts

Gene Symbol
Arhgef40
Entrez Gene ID
268739
Gene Name
Rho guanine nucleotide exchange factor (GEF) 40
Gene Aliases
E130112L23Rik, Gm669, Solo
Chromosome Location
Chr. 14: 51984681 - 52006247 on Build GRCm38
UniGene ID
Mm.9239
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145921.1 NP_001139393.1 17, 18 3940
NM_001145922.1 NP_001139394.1 17, 18 3940
NM_198249.4 NP_937892.2 17, 18 3940
GenBank mRNA AK131136.1 - 17, 18 3909
AK143527.1 - 17, 18 3915
AK163945.1 - 17, 18 4133
BC030925.1 - 4, 5 1099
BC058405.1 - 3, 4 399
BC059018.1 - 17, 18 3842 6848691

View Full Product Details