Gene Transcripts

Gene Symbol
Krtap13-1
Entrez Gene ID
268905
Gene Name
keratin associated protein 13-1
Gene Aliases
AA589563, KAP13.1
Chromosome Location
Chr. 16: 88728862 - 88729611 on Build GRCm38
UniGene ID
Mm.206668
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183189.1 NP_899012.1 1 384
GenBank mRNA BC027551.1 - 1 384

View Full Product Details