Gene Transcripts

Gene Symbol
Ip6k3
Entrez Gene ID
271424
Gene Name
inositol hexaphosphate kinase 3
Gene Aliases
D830007E07Rik, Ihpk3
Chromosome Location
Chr. 17: 27143971 - 27167764 on Build GRCm38
UniGene ID
Mm.209679
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173027.2 NP_766615.1 6 1305
GenBank mRNA AK052857.1 - 6 1306
BC141263.1 - 6 1199 9055875

View Full Product Details