Gene Transcripts

Gene Symbol
Duxbl1
Entrez Gene ID
278672
Gene Name
double homeobox B-like 1
Gene Aliases
1110051B16Rik, Duxbl, Duxl
Chromosome Location
Chr. 14: 25978918 - 25990145 on Build GRCm38
UniGene ID
Mm.428840
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183389.1 NP_899245.1 5 1350
GenBank mRNA AK004227.1 - 1 124
AK141686.1 - 5 1026
BC055782.1 - 5 1350 6403934
EF472598.1 - 5 1087

Gene Symbol
Duxbl2
Entrez Gene ID
100039227
Gene Name
double homeobox B-like 2
Gene Aliases
ENSMUSG00000072675, Gm10394
Chromosome Location
Chr. 14: 26118690 - 26129785 on Build GRCm38
UniGene ID
Mm.428840
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001177538.1 NP_001171009.1 5 1340

Gene Symbol
Duxbl3
Entrez Gene ID
100039293
Gene Name
double homeobox B-like 3
Gene Aliases
ENSMUSG00000072672, Gm10391
Chromosome Location
Chr. 14: 26258303 - 26269434 on Build GRCm38
UniGene ID
Mm.429131
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001177539.1 NP_001171010.1 1340

View Full Product Details