Gene Transcripts

Gene Symbol
Rfx7
Entrez Gene ID
319758
Gene Name
regulatory factor X, 7
Gene Aliases
2510005N23Rik, 9930116O05Rik, D130086K05Rik, Rfxdc2
Chromosome Location
Chr. 9: 72532167 - 72622949 on Build GRCm38
UniGene ID
Mm.32081
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033536.1 NP_001028708.1 9 2459
GenBank mRNA CN537334.1 - 1 533

View Full Product Details