Gene Transcripts

Gene Symbol
Ano4
Entrez Gene ID
320091
Gene Name
anoctamin 4
Gene Aliases
A330096O15Rik, Gm65, Tmem16d
Chromosome Location
Chr. 10: 88948935 - 89345087 on Build GRCm38
UniGene ID
Mm.236464
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001277188.1 NP_001264117.1 23 2716
GenBank mRNA BC065114.1 - 22 2673

View Full Product Details