Gene Transcripts

Gene Symbol
Prr18
Entrez Gene ID
320111
Gene Name
proline rich 18
Gene Aliases
9630019K15Rik
Chromosome Location
Chr. 17: 8340406 - 8344113 on Build GRCm38
UniGene ID
Mm.442510
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178774.4 NP_848889.3 2 1583
GenBank mRNA AK035939.1 - 1 1406
AK039752.1 - 2 1408
AK045379.1 - 2 1717
BC060190.1 - 2 1739 6833275

View Full Product Details