Gene Transcripts

Gene Symbol
Rft1
Entrez Gene ID
328370
Gene Name
RFT1 homolog (S. cerevisiae)
Gene Aliases
D930025H04
Chromosome Location
Chr. 14: 30654341 - 30691317 on Build GRCm38
UniGene ID
Mm.259657
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177815.3 NP_808483.2 1, 2 106
GenBank mRNA AK086390.1 - 1, 2 95
BB652085.1 - 1, 2 106
BC082539.1 - 1, 2 74
BC094336.1 - 1, 2 74
BC116368.2 - 1, 2 87
BC116369.2 - 1, 2 87
BC141275.1 - 1, 2 100 9055887
BC145606.1 - 1, 2 100 9054172

View Full Product Details