Gene Transcripts

Gene Symbol
Sh2d4b
Entrez Gene ID
328381
Gene Name
SH2 domain containing 4B
Gene Aliases
A430109M18Rik, D030001E08
Chromosome Location
Chr. 14: 40811521 - 40902337 on Build GRCm38
UniGene ID
Mm.235694
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177816.3 NP_808484.2 3 454
GenBank mRNA AK020787.1 - 3 760
AX776199.1 - 3 454
BC138964.1 - 3 552 8861986

View Full Product Details