Gene Transcripts
- Gene Symbol
- QARS
- Entrez Gene ID
- 5859
- Gene Name
- glutaminyl-tRNA synthetase
- Gene Aliases
- GLNRS, MSCCA, PRO2195
- Chromosome Location
- Chr. 3: 49095932 - 49105129 on Build GRCh38
- UniGene ID
- Hs.79322
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001272073.1 |
NP_001259002.1
|
21, 22 |
2451 |
|
NM_005051.2 |
NP_005042.1
|
21, 22 |
2484 |
|
NR_073590.1 |
|
21, 22 |
2435 |
|
XM_017006965.1 |
XP_016862454.1
|
21, 22 |
2164 |
|
GenBank mRNA |
AF130067.1 |
- |
17, 18 |
1612 |
|
AK056815.1 |
- |
21, 22 |
2063 |
|
AK222510.1 |
- |
21, 22 |
2112 |
|
AK290523.1 |
- |
21, 22 |
2112 |
|
AK293260.1 |
- |
20, 21 |
2011 |
|
AK297987.1 |
- |
20, 21 |
1958 |
|
AK301559.1 |
- |
21, 22 |
2080 |
|
BC000394.2 |
- |
21, 22 |
2093 |
2821788 |
BC001567.1 |
- |
21, 22 |
2091 |
3452261 |
BC001772.1 |
- |
17, 18 |
1619 |
|
BC016634.1 |
- |
20, 21 |
1916 |
|
BC029739.1 |
- |
21, 22 |
2094 |
3857444 |
X76013.1 |
- |
21, 22 |
2093 |
|
View Full Product Details