Gene Transcripts

Gene Symbol
Rgag4
Entrez Gene ID
331474
Gene Name
retrotransposon gag domain containing 4
Gene Aliases
6430402L03Rik, Mar5, Mart5, mKIAA2001
Chromosome Location
Chr. X: 102066542 - 102071307 on Build GRCm38
UniGene ID
Mm.23297
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278534.1 NP_001265463.1 1 1773
GenBank mRNA AK032151.1 - 1 1663
AK220436.1 - 1 1770
BC132391.1 - 1 1627 40130668
BC137795.1 - 1 1627 8860814

View Full Product Details