Gene Transcripts

Gene Symbol
Cd59b
Entrez Gene ID
333883
Gene Name
CD59b antigen
Gene Aliases
Chromosome Location
Chr. 2: 104069780 - 104085790 on Build GRCm38
UniGene ID
Mm.359026
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181858.1 NP_862906.1 387
GenBank mRNA BC139203.1 - 5 434
BC145912.1 - 5 434 40131215
BC171969.1 - 5 434 9054255

View Full Product Details