Gene Transcripts

Gene Symbol
Prss33
Entrez Gene ID
353130
Gene Name
protease, serine 33
Gene Aliases
Eos, mT6, tryptase-6
Chromosome Location
Chr. 17: 23833338 - 23836126 on Build GRCm38
UniGene ID
Mm.261689
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081399.2 NP_001074868.2 5 667
GenBank mRNA AY262280.1 - 5 643
BC101950.1 - 5 643 40044230
BC103541.1 - 6 739 40044224
BC103542.1 - 5 643 40044219

View Full Product Details