Gene Transcripts

Gene Symbol
RAD17
Entrez Gene ID
5884
Gene Name
RAD17 checkpoint clamp loader component
Gene Aliases
CCYC, HRAD17, R24L, RAD17SP, RAD24
Chromosome Location
Chr. 5: 69369297 - 69414801 on Build GRCh38
UniGene ID
Hs.16184
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278622.1 NP_001265551.1 15 1985
NM_002873.1 NP_002864.1 14 2090
NM_133338.2 NP_579916.1 16 1986
NM_133339.2 NP_579917.1 13 1543
NM_133340.2 NP_579918.1 13 1576
NM_133341.2 NP_579919.1 13 1502
NM_133342.2 NP_579920.1 15 1963
NM_133343.1 NP_579921.1 15 1928
NM_133344.2 NP_579922.1 14 1745
XM_017009679.1 XP_016865168.1 15 1805
XM_017009680.1 XP_016865169.1 14 1859
XM_017009681.1 XP_016865170.1 14 1717
GenBank mRNA AF017748.1 - 14 1715
AF076838.1 - 14 2090
AF085736.2 - 13 1543
AF098533.1 - 13 1522
AF098534.1 - 13 1448
AF112263.1 - 13 1567
AF126424.1 - 16 1985
AJ001642.1 - 14 1754
AJ004977.1 - 14 1750
AK292487.1 - 15 1963
AL122068.1 - 15 1928
BC032304.1 - 13 1556 5170876
BX537441.1 - 15 1985

View Full Product Details