Gene Transcripts
- Gene Symbol
- Slirp
- Entrez Gene ID
- 380773
- Gene Name
- SRA stem-loop interacting RNA binding protein
- Gene Aliases
- 1810035L17Rik
- Chromosome Location
- Chr. 12: 87443896 - 87449924 on Build GRCm38
- UniGene ID
- Mm.355701
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_026958.3 |
NP_081234.2
|
1, 2 |
137 |
|
GenBank mRNA |
AJ293625.1 |
- |
1, 2 |
106 |
|
AK007699.1 |
- |
1, 2 |
124 |
|
AK028746.1 |
- |
|
115 |
|
BC048513.1 |
- |
1, 2 |
117 |
|
BC065396.1 |
- |
1, 2 |
106 |
|
BC100567.1 |
- |
1, 2 |
102 |
|
BC116739.1 |
- |
1, 2 |
124 |
40126058 |
BC116741.1 |
- |
1, 2 |
124 |
40126060 |
BU850988.1 |
- |
1, 2 |
140 |
|
CK372908.1 |
- |
1, 2 |
107 |
|
View Full Product Details