Gene Transcripts

Gene Symbol
Spag6l
Entrez Gene ID
381350
Gene Name
sperm associated antigen 6 like
Gene Aliases
Chromosome Location
Chr. 2: 18694011 - 18749746 on Build GRCm38
UniGene ID
Mm.330024
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001334.2 NP_001001334.2 8, 9 1239
GenBank mRNA AK132882.1 - 8, 9 1239
BC049634.1 - 4, 5 423
BC061194.1 - 8, 9 1236

View Full Product Details