Gene Transcripts

Gene Symbol
Rxfp1
Entrez Gene ID
381489
Gene Name
relaxin/insulin-like family peptide receptor 1
Gene Aliases
Gm1018, Lgr7
Chromosome Location
Chr. 3: 79641600 - 79737951 on Build GRCm38
UniGene ID
Mm.331086
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_212452.1 NP_997617.1 6, 7 545
GenBank mRNA AK137655.1 - 5, 6 526
AY509975.1 - 6, 7 545

View Full Product Details