Gene Transcripts

Gene Symbol
Mup21
Entrez Gene ID
381531
Gene Name
major urinary protein 21
Gene Aliases
Gm11208, Mup, Mup26, OTTMUSG00000000231, bM64F17.1, bM64F17.4
Chromosome Location
Chr. 4: 62147832 - 62150841 on Build GRCm38
UniGene ID
Mm.472122
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009550.2 NP_001009550.1 4 460
GenBank mRNA BC141241.1 - 4 455
EU882236.1 - 4 371

View Full Product Details