Gene Transcripts

Gene Symbol
Thap6
Entrez Gene ID
381650
Gene Name
THAP domain containing 6
Gene Aliases
4930534K21Rik, AI448650
Chromosome Location
Chr. 5: 91962882 - 91972137 on Build GRCm38
UniGene ID
Mm.159453
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK015967.1 - 4 1057
AK039179.1 - 2 696
AK080650.1 - 2 695
BC089377.1 - 3 369

View Full Product Details