Gene Transcripts

Gene Symbol
Ppapdc1a
Entrez Gene ID
381925
Gene Name
phosphatidic acid phosphatase type 2 domain containing 1A
Gene Aliases
C030048B12Rik, Gm1090
Chromosome Location
Chr. 7: 129257094 - 129391307 on Build GRCm38
UniGene ID
Mm.39184
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080963.1 NP_001074432.1 4 318
GenBank mRNA AK081292.1 - 4 383
BC120497.1 - 4 318 40129420

View Full Product Details