Gene Transcripts

Gene Symbol
Fbxw15
Entrez Gene ID
382105
Gene Name
F-box and WD-40 domain protein 15
Gene Aliases
Fbxo12J
Chromosome Location
Chr. 9: 109552602 - 109569438 on Build GRCm38
UniGene ID
Mm.335468
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199036.2 NP_950201.2 7 877
GenBank mRNA AK087669.1 - 7 878
DQ067445.1 - 7 845

View Full Product Details