Gene Transcripts

Gene Symbol
Fbxw26
Entrez Gene ID
382109
Gene Name
F-box and WD-40 domain protein 26
Gene Aliases
EG382109, Gm5163
Chromosome Location
Chr. 9: 109717566 - 109746089 on Build GRCm38
UniGene ID
Mm.246849
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198674.2 NP_941076.1 3 183
GenBank mRNA AK054298.1 - 3 183
BC132114.1 - 3 182 40130391
BC132304.1 - 3 182 40130581

View Full Product Details