Gene Transcripts
- Gene Symbol
- Sly
- Entrez Gene ID
- 382301
- Gene Name
- Sycp3 like Y-linked
- Gene Aliases
- Chromosome Location
- Chr. Y: 57187206 - 57213358 on Build GRCm38
- UniGene ID
- Mm.304354
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_201530.2 |
NP_963288.2
|
4, 5 |
273 |
|
GenBank mRNA |
BC049626.1 |
- |
4, 5 |
273 |
|
View Full Product Details