Gene Transcripts

Gene Symbol
Kcnf1
Entrez Gene ID
382571
Gene Name
potassium voltage-gated channel, subfamily F, member 1
Gene Aliases
Gm1182, Kv5.1
Chromosome Location
Chr. 12: 17172100 - 17176888 on Build GRCm38
UniGene ID
Mm.40173
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_201531.3 NP_963289.2 1 1446
GenBank mRNA AK137367.1 - 1 1139
BC053089.1 - 1 1083 5709192

View Full Product Details