Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006909.2 | NP_008840.1 | 18, 19 | 2851 | |
XM_017009682.1 | XP_016865171.1 | 18, 19 | 2533 | ||
GenBank mRNA | AF023130.1 | - | 18, 19 | 2801 | |
AF181250.1 | - | 18, 19 | 2801 | ||
BC126112.1 | - | 18, 19 | 2851 | 8991828 | |
BC136296.1 | - | 18, 19 | 2851 | 9020283 | |
BC143870.1 | - | 18, 19 | 2851 |