Gene Transcripts

Gene Symbol
Fndc5
Entrez Gene ID
384061
Gene Name
fibronectin type III domain containing 5
Gene Aliases
1500001L03Rik, AI836596, C87088, PeP, Pxp
Chromosome Location
Chr. 4: 129136922 - 129144593 on Build GRCm38
UniGene ID
Mm.44075
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027402.3 NP_081678.1 3 314
GenBank mRNA AJ401376.1 - 3 314
AK005096.1 - 3 376
BC109183.1 - 3 310 40050064
BC109184.1 - 3 314 40050065

View Full Product Details