Gene Transcripts

Gene Symbol
Adam20
Entrez Gene ID
384806
Gene Name
a disintegrin and metallopeptidase domain 20
Gene Aliases
4930529F22Rik, Adam38
Chromosome Location
Chr. 8: 40793273 - 40797342 on Build GRCm38
UniGene ID
Mm.444736
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009548.2 NP_001009548.1 2 2314
GenBank mRNA AK133113.1 - 2 2315
BC150793.1 - 2200 9087704

View Full Product Details