Gene Transcripts

Gene Symbol
Otud6a
Entrez Gene ID
408193
Gene Name
OTU domain containing 6A
Gene Aliases
EG408193, Hshin6
Chromosome Location
Chr. X: 100429013 - 100429885 on Build GRCm38
UniGene ID
Mm.469910
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163191.1 NP_001156663.1 1 256
GenBank mRNA BC153817.1 - 1 256

View Full Product Details