Gene Transcripts

Gene Symbol
Mfsd2b
Entrez Gene ID
432628
Gene Name
major facilitator superfamily domain containing 2B
Gene Aliases
Gm1964
Chromosome Location
Chr. 12: 4862438 - 4875044 on Build GRCm38
UniGene ID
Mm.154078
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033488.2 NP_001028660.2 6, 7 676
GenBank mRNA AK133061.1 - 6, 7 691
AK156138.1 - 7, 8 828
AK157892.1 - 5, 6 1503
AK162620.1 - 6, 7 675
AK172423.1 - 6, 7 677
BC145584.1 - 6, 7 689

View Full Product Details