Gene Transcripts

Gene Symbol
Rhox8
Entrez Gene ID
434768
Gene Name
reproductive homeobox 8
Gene Aliases
Tox
Chromosome Location
Chr. X: 37874776 - 37890766 on Build GRCm38
UniGene ID
Mm.336025
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004193.2 NP_001004193.1 3 962
GenBank mRNA AY329370.1 - 3 962
BC139098.1 - 3 945 8862120
DQ058645.1 - 4 971

View Full Product Details