Gene Transcripts

Gene Symbol
Taar7f
Entrez Gene ID
435207
Gene Name
trace amine-associated receptor 7F
Gene Aliases
taR-7f
Chromosome Location
Chr. 10: 24049510 - 24050586 on Build GRCm38
UniGene ID
Mm.377966
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001010839.1 NP_001010839.1 1 947
GenBank mRNA BC139175.1 - 1 975
BC139176.1 - 1 975 8862198

View Full Product Details