Gene Transcripts

Gene Symbol
Cyp4a30b
Entrez Gene ID
435802
Gene Name
cytochrome P450, family 4, subfamily a, polypeptide 30b
Gene Aliases
Cyp4a30b-ps, OTTMUSG00000008626
Chromosome Location
Chr. 4: 115452491 - 115471418 on Build GRCm38
UniGene ID
Mm.466611
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100185.1 NP_001093655.1 4 422

View Full Product Details