Gene Transcripts

Gene Symbol
Baiap3
Entrez Gene ID
545192
Gene Name
BAI1-associated protein 3
Gene Aliases
Bap3, Gm937
Chromosome Location
Chr. 17: 25242659 - 25256364 on Build GRCm38
UniGene ID
Mm.328859
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163270.1 NP_001156742.1 31 3155
GenBank mRNA AK122358.1 - 29 3274
BC158116.1 - 31 3155 9088189

View Full Product Details