Gene Transcripts

Gene Symbol
Cyp2c67
Entrez Gene ID
545288
Gene Name
cytochrome P450, family 2, subfamily c, polypeptide 67
Gene Aliases
C730004C24Rik
Chromosome Location
Chr. 19: 39608844 - 39649083 on Build GRCm38
UniGene ID
Mm.297048
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024719.2 NP_001019890.1 1, 2 182
GenBank mRNA AK149389.1 - 1, 2 193
BC092260.1 - 1, 2 182 5132591
BC106142.1 - 1, 2 182 5051946

Gene Symbol
Cyp2c40
Entrez Gene ID
13099
Gene Name
cytochrome P450, family 2, subfamily c, polypeptide 40
Gene Aliases
AI265721, AI662255
Chromosome Location
Chr. 19: 39767073 - 39812814 on Build GRCm38
UniGene ID
Mm.353875
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010004.2 NP_034134.2 1, 2 262
GenBank mRNA AF047727.1 - 1, 2 176
BC139471.1 - 1, 2 262 8862493
BC139472.1 - 1, 2 262 8862494

Gene Symbol
Cyp2c69
Entrez Gene ID
100043108
Gene Name
cytochrome P450, family 2, subfamily c, polypeptide 69
Gene Aliases
AI098658
Chromosome Location
Chr. 19: 39842660 - 39886769 on Build GRCm38
UniGene ID
Mm.482331
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001104525.1 NP_001097995.1 1, 2 232
GenBank mRNA BC145630.1 - 1, 2 262

View Full Product Details