Gene Transcripts

Gene Symbol
Smok3a
Entrez Gene ID
545814
Gene Name
sperm motility kinase 3A
Gene Aliases
Smok2b1
Chromosome Location
Chr. 5: 138021276 - 138034665 on Build GRCm38
UniGene ID
Mm.349005
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001126045.1 NP_001119517.1 2 817
NM_001168602.1 NP_001162073.1 2 970
GenBank mRNA AJ245454.1 - 2 970
BC150960.1 - 817 9087871

Gene Symbol
Smok3b
Entrez Gene ID
622474
Gene Name
sperm motility kinase 3B
Gene Aliases
4931427P12Rik, Smok2b2
Chromosome Location
Chr. 5: 138037224 - 138050633 on Build GRCm38
UniGene ID
Mm.456691
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039889.3 NP_001034978.1 2 991
GenBank mRNA AK133180.1 - 2 992

View Full Product Details