Gene Transcripts

Gene Symbol
Zscan4d
Entrez Gene ID
545913
Gene Name
zinc finger and SCAN domain containing 4D
Gene Aliases
EG545913
Chromosome Location
Chr. 7: 11160883 - 11167460 on Build GRCm38
UniGene ID
Mm.380086
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100186.1 NP_001093656.1 2 250
GenBank mRNA EF143406.1 - 2 250

View Full Product Details