Gene Transcripts

Gene Symbol
Psg27
Entrez Gene ID
545925
Gene Name
pregnancy-specific glycoprotein 27
Gene Aliases
EG545925, cea15
Chromosome Location
Chr. 7: 18556514 - 18567305 on Build GRCm38
UniGene ID
Mm.440302
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037168.1 NP_001032245.1 4 1197
GenBank mRNA BC100351.1 - 4 1197

View Full Product Details