Gene Transcripts

Gene Symbol
RPL9
Entrez Gene ID
6133
Gene Name
ribosomal protein L9
Gene Aliases
L9, NPC-A-16
Chromosome Location
Chr. 4: 39454124 - 39458948 on Build GRCh38
UniGene ID
Hs.412370
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000661.4 NP_000652.2 6 487
NM_001024921.3 NP_001020092.1 6 645
GenBank mRNA AB062431.1 - 1 434
AK130966.1 - 1 450
AK312241.1 - 1 454
AL832047.1 - 3 3522
AY320412.1 - 6 470
AY376242.1 - 5 604
AY597415.1 - 1 461
BC000483.2 - 1 439 2964733
BC004156.2 - 1 433 2961026
BC004206.2 - 1 433 2961026
BC007967.2 - 1 489 4304670
BC012149.1 - 1 489 4554838
BC031906.1 - 5 434 4996986
BC066318.1 - 1 454 5277491
BC070214.1 - 1 493 6605055
BC100285.1 - 5 594 6022858
BG829769.1 - 5 435
D14531.1 - 5 434
U09953.1 - 6 457
U21138.1 - 1 434

View Full Product Details